Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRIM29 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-B16S 600-401-B16S
RL600-401-B16SEA 148 USD InStock
RL600-401-B16S CA600-401-B16S
Anti-TRIM29 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chimpanzee, Horse, Human, macaque

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty