Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SAE1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-B24 600-401-B24S
RL600-401-B24EA 518 USD InStock
RL600-401-B24 RL600-401-B24S
Anti-SAE1 Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~37 kDa in size corresponding to phosphorylated SAE1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 2µg/ml;

Type: Primary
Antigen: SAE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Mouse, Rat, Dog

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty