Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASZ1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-B62 600-401-B62S
RL600-401-B62EA 518 USD InStock
RL600-401-B62 RL600-401-B62S
Anti-CASZ1 Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA and western blotting. This antiserum detects endogenous Casz1 Proteins, both the hCasz5 and hCasz11 isoforms. Expect a band approximately 125 kDa for hCasz5 and 190 kDa for hCaz11.

Recommended Dilutions: ELISA: 1:45000; Western Blot: 1:1000 - 1:5000;

Type: Primary
Antigen: CASZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Drosophila, Chimpanzee, Macaque

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty