Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RRM2B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-B67 600-401-B67S
RL600-401-B67EA 518 USD InStock
RL600-401-B67 RL600-401-B67S
Anti-RRM2B Rabbit Polyclonal Antibody
Antibodies
RRM2B/p53R2 antibody is suitable for IP, IHC, IF, ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 41 kDa in size corresponding to RRM2B1 by western blotting.

Recommended Dilutions: ELISA: 1:3000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: RRM2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty