Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RTN4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-C01
RL600-401-C01EA 518 USD InStock
RL600-401-C01
Anti-RTN4 Rabbit Polyclonal Antibody
Antibodies
Anti-NogoA antibody has been tested for use in ELISA, Western Blot, and in immunochemistry. Specific conditions for reactivity should be optimized by the end user. Despite its predicted molecular weight, NogoA typically migrates at 180kDa in an SDS-PAGE.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 0.5 - 1.0 µg/ml; Immunochemistry: 2.5 µg/ml;

Type: Primary
Antigen: RTN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty