Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-C64S 600-401-C64
RL600-401-C64SEA 148 USD InStock
RL600-401-C64S RL600-401-C64
Anti-STAT3 Rabbit Polyclonal Antibody
Antibodies
This affinity purified Stat3 antibody has been tested for use in ELISA, IHC, and Western Blot. Expect a band approximately 88.1 kDa in size corresponding to phosphorylated Stat3 Protein by western blotting in the appropriate stimulated tissue or cell lysate.

Recommended Dilutions: ELISA: 1:60,000-1:130,000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty