Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIN2B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-C87
RL600-401-C87EA 518 USD InStock
RL600-401-C87
Anti-GRIN2B Rabbit Polyclonal Antibody
Antibodies
Anti-NMDA NR2B subunit (Rabbit) antibody is suitable for use in Western Blotting, Immunoprecipitation and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band or approximately 180 kDa in size corresponding to the NR2B subunit.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000-1:2000; Immunoprecipitation: 3µl per 200µg lysate;

Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty