Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EPHB2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-D36
RL600-401-D36EA 518 USD InStock
RL600-401-D36
Anti-EPHB2 Rabbit Polyclonal Antibody
Antibodies
Anti-EphrinB pT317 Antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 46 kDa in size corresponding to EphrinB Protein phosphorylated at Tyr317.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: EPHB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Chicken

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty