Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-D85
RL600-401-D85EA 518 USD InStock
RL600-401-D85
Anti-GRIN1 Rabbit Polyclonal Antibody
Antibodies
Anti-NMDA R1, Splice Variant C1 (Rabbit) Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 120 kDa in size corresponding to the NR1 subunit.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000-2000;
0.05 M Tris Chloride, 0.15 M Sodium Chloride, pH 7.4;

Type: Primary
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty