Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POSTN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-E10 600-401-E09
RL600-401-E09EA 518 USD InStock
RL600-401-E09 RL600-401-E10
Anti-POSTN Rabbit Polyclonal Antibody
Antibodies
Anti-Periostin Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a three bands of approximately 93 kDa in size corresponding to periostin pan Proteins in mouse lung extract.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100;

Type: Primary
Antigen: POSTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Chicken, Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty