Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAOK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-E41
RL600-401-E41EA 518 USD InStock
RL600-401-E41
Anti-TAOK2 Rabbit Polyclonal Antibody
Antibodies
Anti-TAO2 pS181 (Rabbit) antibody is suitable for use in Western Blotting. Expect a band of approximately 120 kDa in size corresponding to TAO2 Protein phosphorylated at Ser181 in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TAOK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty