Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TH Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-E51
RL600-401-E51EA 518 USD InStock
RL600-401-E51
Anti-TH Rabbit Polyclonal Antibody
Antibodies
Anti-Tyrosine Hydroxylase pS31 (Rabbit) antibody is suitable for use in Western Blotting, Immunochemistryand IF. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 60 kDa in size corresponding to tyrosine hydroxylase.

Recommended Dilutions: Immunofluorescence: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:1000;

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty