Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BSN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-E93
RL600-401-E93EA 474 USD InStock
RL600-401-E93
Anti-BSN Rabbit Polyclonal Antibody
Antibodies
Anti-Bassoon Antibody is suitable for use in WB and IHC. Expect a band approximately ~420kDa on specific lysates corresponding to the molecular mass of Bassoon.

Recommended Dilutions: Western Blot: 1:1000;

Type: Primary
Antigen: BSN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty