Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPH1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-F52
RL600-401-F52EA 491 USD InStock
RL600-401-F52
Anti-HSPH1 Rabbit Polyclonal Antibody
Antibodies
Anti-Hsp110 Antibody is suitable for use in WB. Expect a band approximately ~110kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HSPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Hamster, Rat, Mouse, Bovine, Sheep, Yeast, Armenian Hamster

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty