Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IDO1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J43 600-401-J43
RL600-401-J43EA 518 USD InStock
RL600-401-J43 CA10037-254
Anti-IDO1 Rabbit Polyclonal Antibody
Antibodies
Anti-IDO antibody is useful for ELISA, immunohistochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~41kDa corresponding to the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1µg/ml

Type: Primary
Antigen: IDO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty