Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KMO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J84 600-401-J84
RL600-401-J84EA 518 USD InStock
CA10037-336 RL600-401-J84
Anti-KMO Rabbit Polyclonal Antibody
Antibodies
Anti-KMO antibody is useful for ELISA, Immunohistochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~58kDa corresponding to the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1µg/ml

Type: Primary
Antigen: KMO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty