Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIST2H3C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-K01 600-401-K01
RL600-401-K01EA 518 USD InStock
RL600-401-K01 CA10037-362
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antibodies
Anti-Histone H3 (Dimethyl Lys4/pT6) antibody is useful for ELISA, immunochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~15.4kDa corresponding to the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000; Immunofluorescence: 1:50; Western Blot: 1:500; Immunochemistry: 1:50; Immunoprecipitation: 1:100

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, C. elegans

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty