Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIST2H3C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P08 600-401-P08
RL600-401-P08EA 539 USD InStock
RL600-401-P08 CA600-401-P08
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antibodies
Anti-Histone H3 K9-Ac/pS10 antibody is useful for Western Blot and Dot Blot.

Recommended Dilutions: Immunofluorescence: 1:10; Western Blot: 1.0 µg/ml

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty