Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H3F3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P10 600-401-P10
RL600-401-P10EA 539 USD InStock
RL600-401-P10 CA600-401-P10
Anti-H3F3A Rabbit Polyclonal Antibody
Antibodies
Anti-Histone H3 K79-Me1 antibody is has been tested for use in Western Blot, ChIP, Immunofluorescence, and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~15kDa.

Recommended Dilutions: ChIP: 2 µg; Immunofluorescence: 1:50 - 1:100; Western Blot: 0.5µg/ml

Type: Primary
Antigen: Histone H3 (HIST2H3C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: C. elegans, Drosophila, Human, Mouse, Plant, Primate, Rat, Xenopus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty