Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Biotin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
600-403-098
RL600-403-098EA 430 USD InStock
RL600-403-098
Anti-Biotin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:10000-1:80,000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty