Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

600-403-261 600-403-261
RL600-403-261EA 539 USD InStock
CARL600403-261 RL600-403-261
Anti-S Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
HRP Conjugated Anti-Phosphoserine antibody is suitable for use in ELISA and western blotting assays.  The antibody reacts specifically with phosphoserine and shows minimal reactivity by ELISA and western blotting with Proteins.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:500;
Do not add Sodium Azide!

Type: Primary
Antigen: S
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty