Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-T Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
600-403-263 600-403-263
RL600-403-263EA 539 USD InStock
CA600-403-263 RL600-403-263
Anti-T Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Phospho Site Specific Threonine affinity purified antibody is suitable for use in ELISA and western blotting assays.  The antibody reacts specifically with phosphothreonine and shows minimal reactivity by ELISA and western blotting with Proteins.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:500;

Type: Primary
Antigen: T
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty