Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ALB Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-033
RL600-406-033EA 1320 USD InStock
RL600-406-033
Anti-ALB Rabbit Polyclonal Antibody (Biotin)
Antibodies
Anti-HUMAN SERUM ALBUMIN (RABBIT) Antibody Biotin Conjugated antibody has been tested in ELISA, western blot, and immunohistochemical assays. Optimal concentrations should be determined by researcher.

Recommended Dilutions: ELISA: 1:60,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Primary
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty