Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COL5A3 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-107 600-406-107
RL600-406-107EA 539 USD InStock
RL600-406-107 CA600-406-107
Anti-COL5A3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
This product was assayed by immunoblot and found to be reactive against Collagen V at a dilution of 1:5,000 to 1:10,000.

Recommended Dilutions: ELISA: 1:4000 to 1:8000; Western Blot: 1:5000 to 1:10000; Immunochemistry: 1:50 to 1:200; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: COL5A3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty