Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COL6A1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-108 600-406-108
RL600-406-108EA 539 USD InStock
RL600-406-108 CA600-406-108
Anti-COL6A1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Anti-Collagen Type VI (RABBIT) Antibody Biotin Conjugated is suitable for western blot, immunoprecipitatioin, and immunochemistry. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: COL6A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty