Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAMB1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-116 600-406-116
RL600-406-116EA 508 USD InStock
CA600-406-116 RL600-406-116
Anti-LAMB1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Anti-Laminin biotin conjugated antibody is suitable for western blotting, IHC, IP and for ELISA and for use with streptavidin reporter reagents. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: LAMB1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty