Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLH Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-466
RL600-406-466EA 487 USD InStock
RL600-406-466
Anti-KLH Rabbit Polyclonal Antibody (Biotin)
Antibodies
Anti-KLH Antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Anti-KLH antibody biotin conjugate is suitable for use with streptavidin reporter systems.

Recommended Dilutions: ELISA: 1:250000; Western Blot: 1:5000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty