Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AcK Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-939 600-406-939
RL600-406-939EA 538 USD InStock
RL600-406-939 CARL600406-939
Anti-AcK Rabbit Polyclonal Antibody (Biotin)
Antibodies
This affinity-purified antibody is suitable for use in ELISA and western blotting assays.  The antibody reacts specifically with acetylated lysine residues.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000-1:5000;

Type: Primary
Antigen: AcK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Mammalian

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty