Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-V5 Rabbit Polyclonal Antibody (DyLight® 649)

Supplier: Rockland Immunochemicals
600-443-378 600-443-378
RL600-443-378EA 487 USD InStock
CA600-443-378 RL600-443-378
Anti-V5 Rabbit Polyclonal Antibody (DyLight® 649)
Antibodies
Primary Rabbit Anti-V5 EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: V5
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope:
Host: Rabbit
Isotype:
Reactivity: Many Species

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty