Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-6xHis Rabbit Polyclonal Antibody (DyLight® 800)

Supplier: Rockland Immunochemicals
600-445-382 600-445-382
RL600-445-382EA 454 USD InStock
CA600-445-382 RL600-445-382
Anti-6xHis Rabbit Polyclonal Antibody (DyLight® 800)
Antibodies
Primary Rabbit Anti-6X HIS EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty