Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Sheep Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-601-401 600-601-401
RL600-601-401EA 518 USD InStock
CA600-601-401 RL600-601-401
Anti-AKT1 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Sheep
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty