Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DBH Sheep Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-601-D28 600-601-D27
RL600-601-D27EA 518 USD InStock
RL600-601-D27 RL600-601-D28
Anti-DBH Sheep Polyclonal Antibody
Antibodies
Anti-Dopamine beta-Hydroxylase Antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optmized by the end user. Expect a band of approximately 75 kDa in size corresponding to DBH Protein.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: DBH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Sheep
Isotype: IgG
Reactivity: Human, non-Human Primate

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty