Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-901-215S 600-901-215S
RL600-901-215SEA 150 USD InStock
CA600-901-215S RL600-901-215S
Anti-GFP Chicken Polyclonal Antibody
Antibodies
Anti-GFP is designed to detect GFP and its variants. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen and recognizes wild type, recombinant and enhanced forms of GFP.

Recommended Dilutions: ELISA: 1:2000 - 1:12000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:200 - 1:1000;

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Chicken
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty