Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ctnnb1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-C68S 600-401-C68
RL600401C68SEA 148 USD InStock
RL600401C68S RL600401C68
Anti-ctnnb1 Rabbit Polyclonal Antibody
Antibodies
beta catenin antibody has been tested for use in ELISA, western blotting, and ISH. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 85.5 kDa in size.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:500; Other Dilution: ISH

Type: Primary
Antigen: ctnnb1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty