Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPRY2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-E68 600-401-E68S
RL600401E68SEA 148 USD InStock
RL600401E68S RL600401E68
Anti-SPRY2 Rabbit Polyclonal Antibody
Antibodies
Anti-Sprouty-2 antibody is suitable for ELISA, Immunofluorescence, Immunohistochemistry, and western blotting. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: User Optimized; Immunofluorescence: User Optimized; Western Blot: 1:500-1:5000; Immunochemistry: 10 µg/ml

Type: Primary
Antigen: SPRY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty