Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Monoclonal Antibody [clone: 9F9.F9] (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Rockland Immunochemicals
602-1004 602-1004
RL602-1004EA 177 USD InStock
CA602-1004 RL602-1004
Anti-IgG Goat Monoclonal Antibody [clone: 9F9.F9] (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Secondary Goat Anti-IgG F(ab')2 Reacts with Cat (Feline)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Cat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty