Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Serum Proteins IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
605-403-B69
RL605-403-B69EA 150 USD InStock
RL605-403-B69
Anti-Serum Proteins IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended Dilutions: ELISA: 1:120000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:1000 - 1:5000
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Recommended Dilutions: ELISA: 1:120000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:1000 - 1:5000
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat, Mouse, Rat , Human Serum Proteins

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty