Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG gamma Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Rockland Immunochemicals
609-1012 609-1012
RL609-1012EA 216 USD InStock
CA609-1012 RL609-1012
Anti-IgG gamma Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Secondary Goat Anti-IgG (gamma chain) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: gamma chain
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty