Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
609-1103 609-1103
RL609-1103EA 258 USD InStock
CA609-1103 RL609-1103
Anti-IgG Goat Polyclonal Antibody
Antibodies
Human IgG F(c) Antibody is suitable for immunoassays where specificity to the immunoglobulin Fc region is desired. Antibody has been tested by ELISA, western blot immunoblot, and immunohistochemisty.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:10000 - 1:50000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty