Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgL Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
609-1611 609-1611
RL609-1611EA 271 USD InStock
CA609-1611 RL609-1611
Anti-IgL Goat Polyclonal Antibody (Biotin)
Antibodies
Secondary Goat Anti-λ (lambda chain) Reacts with Human

Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty