Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Mouse Polyclonal Antibody

Supplier: Rockland Immunochemicals
609-301-002 609-301-002
RL609-301-002EA 179 USD InStock
CA609-301-002 RL609-301-002
Anti-IgG Mouse Polyclonal Antibody
Antibodies
Secondary Mouse Anti-IgG (H&L) Reacts with Human

Recommended Dilutions: ELISA: 1:5000 - 1:50000; Western Blot: 1:500 - 1:5000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty