Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgA alpha Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
609-4106 609-4106
RL609-4106EA 177 USD InStock
RL609-4106 CA609-4106
Anti-IgA alpha Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti-IgA (alpha chain) Reacts with Human

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:250 - 1:1000;

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: alpha chain
Host: Rabbit
Isotype: IgA
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty