Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgA/IgG/IgM Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
610-101-430
RL610-101-430EA 189 USD InStock
RL610-101-430
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000

Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Bovine, Horse, Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty