Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG2b gamma Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
610-109-042
RL610-109-042EA 243 USD InStock
RL610-109-042
Anti-IgG2b gamma Goat Polyclonal Antibody (Texas Red®)
Antibodies
Anti-Mouse IgG2b Texas Red® Conjugated antibodies are suitable for ELISA, immunochemistry, western blotting, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Western Blot: 1:4000 - 1:20000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent);

Type: Secondary
Antigen: IgG2b gamma
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty