Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG3 gamma Rabbit Polyclonal Antibody (DyLight® 800)

Supplier: Rockland Immunochemicals
610-445-043 610-445-043
RL610-445-043EA 130 USD InStock
CA610-445-043 RL610-445-043
Anti-IgG3 gamma Rabbit Polyclonal Antibody (DyLight® 800)
Antibodies
Type: Secondary
Antigen: IgG3 gamma
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty