Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ig kappa Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
610-4610 610-4610
RL610-4610EA 274 USD InStock
RL610-4610 CA610-4610
Anti-Ig kappa Rabbit Polyclonal Antibody (Biotin)
Antibodies
Secondary Rabbit Anti-κ (kappa chain) Reacts with Mouse

Recommended Dilutions: ELISA: 1:600,000; Western Blot: 1:10000 - 1:50000; Immunochemistry: 1:2000 - 1:10000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: Igkappa
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty