Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgL Rabbit Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
610-4911 610-4911
RL610-4911EA 274 USD InStock
RL610-4911 CA610-4911
Anti-IgL Rabbit Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Rabbit Anti-λ (lambda chain) Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty