Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Donkey Polyclonal Antibody (Cy2®)

Supplier: Rockland Immunochemicals
610-711-124 610-711-124
RL610-711-124EA 302 USD InStock
CA610-711-124 RL610-711-124
Anti-IgG Donkey Polyclonal Antibody (Cy2®)
Antibodies
Secondary Donkey Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions: ELISA: 1:10000 - 1:50000; FLISA: 1:10000 - 1:50000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty