Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Rockland Immunochemicals
610-1002-0500
RL6101002-0500EA 87 USD InStock
RL6101002-0500
Anti-IgG Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500; Other Dilution: FLOW CYTOMETRY 1:500 - 1:2500

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty