Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
610-101-042 610-101-041 610-101-040
RL610101042EA 210 USD InStock
RL610101042 RL610101041 RL610101040
Anti-IgG1 Goat Polyclonal Antibody
Antibodies
Anti-Mouse IgG1 secondary antibody is suitable for ELISA, Immunohistochemistry western blotting as well as other anti IgG1 antibody based assays.

Recommended Dilutions: ELISA: 1:40000 - 1:200000; Western Blot: 1:4000 - 1:20000; Immunochemistry: 1:2000 - 1:10000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty